Learn More
Lactose, Serva Electrophoresis™
Specifica
Specifica
| CAS | 5965-66-2 |
| Formula molecolare | C12H22O11 |
| Numero MDL | MFCD00064521 |
| Quantità | 1 kg |
| Sinonimo | beta-lactose, beta-d-lactose, lactose, fast-flo lactose, 4-o-beta-d-galactopyranosyl-beta-d-glucopyranose, milk sugar, d-lactose, fast-flo, lactose anhydrous, pharmatose 450m |
| InChI Key | GUBGYTABKSRVRQ-DCSYEGIMSA-N |
| SMILES | OC[C@H]1O[C@@H](O[C@H]2[C@H](O)[C@@H](O)[C@H](O)O[C@@H]2CO)[C@H](O)[C@@H](O)[C@H]1O |
| IUPAC Name | (2R,3R,4S,5R,6S)-2-(hydroxymethyl)-6-{[(2R,3S,4R,5R,6R)-4,5,6-trihydroxy-2-(hydroxymethyl)oxan-3-yl]oxy}oxane-3,4,5-triol |
| Molecular Weight (g/mol) | 342.30 |
| PubChem CID | 6134 |
| Vedi altri risultati |