Learn More
Uvex™ Ladies Trousers 7429-Midnight Navy
Catalog Number 17246971
Click to view available options
Size:
19
20
21
22
23
24
25
26
34
36
38
40
42
44
46
48
50
52
54
The uvex suXXeed collection gets everything spot on: well thought-out functionality, sporty design, exciting color contrasts, unusual material combinations and a cut that fits like a glove. Can be worn in industrial and metal processing settings as well as for leisure.
  • Material: 65 % polyester, 35 % cotton 215 g/m², twill weave
  • Comfortable ladies' trousers with elastic.
  • Waistband with elastic and in the front using smooth waistband
  • 1 side pouch pocket in the right side seam, closure with an concealed press stud, suitable processing for industrial laundries
  • Oeko-Tex Standard 100
  • Sizes: 34 - 54
  • Care instructions: 60°C washable, chlorinate impossible, ironing: II points, dry cleaning: P, tumble dryer: I point suitable for industrial laundering
Size 50
Sesso Women's
Colore Blue
Tipo di chiusura Buttons; Zipper
Monouso No
Spessore appro x 0.245 g/m²