Learn More
Corning™ Lambda™ Plus 8-Channels Pipettor, Calibration 3x4 ALT
Codice prodotto. 17203514 Sfoglia Tutto Corning Prodotti
Cambia vista
Click to view available options
Volume (sistema metrico):
μ
5 to 50 μL
50 to 300 μL
Da 20 a 200 μl
Dimensione della confezione:
Pezzo
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Volume (sistema metrico) unitSize
17203514 μ Pezzo
17223514 5 to 50 μL Pezzo
17243514 Da 20 a 200 μl Pezzo
17263514 50 to 300 μL Pezzo
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Codice prodotto. 17203514 Fornitore Corning™ N. del fornitore 4080ALT
Questo articolo non è attualmente disponibile o è stato dismesso.
La preghiamo di consultare la pagina del prodotto per individuare le possibili alternative.
Visualizzare prodotti alternativi
Engineered to provide the highest levels of accuracy, reproducibility, and comfort.
Lambda Plus pipettors feature smooth plunger movement and extremely low pipetting forces. Volume can be set by turning either the thumb wheel or the plunger button, and a locking mechanism prevents accidental change. The Lambda Plus pipettors have been designed to maintain their calibration, even after repeated use and autoclaving. A key is included with each pipettor for easy in-house calibration.
  • Pre-Calibrated Pipettor delivered with ISO 17025 Certificate
  • Test Procedure: 4 measurements at 3 volumes (nominal volume, 50% of nominal volume, 10% of nominal volume or minimal volume)
  • Factory calibrated to strict ISO 8655 standards-3-year warranty
  • Fully autoclavable and UV resistant
  • Designed for universal fit with all common brands of pipet tips
  • Adjustable tip ejector
  • Accuracy (%) ±12 to ±1.5
  • Precision (%) <6.0 to <0.7
TRUSTED_SUSTAINABILITY
Specifica
Certificazione/Conformità ISO17025
Autoclavabile Autoclavable
Da utilizzare con Universal Pipette Tip
Numero di canali 8
Product Line Lambda™ Plus
Tipo Multi-Channel Pipette
Volume (sistema metrico) μ
Monitoraggio UV
Warranty 3 Years
Titolo del prodotto
Wählen Sie ein Thema
Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.