Learn More
Zwiebel™ Large Dust Brush
Product Code. 11781536
Change view
Click to view available options
Length (Metric):
190 mm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Unit Size
11781536 190 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11781536 Supplier Zwiebel™ Supplier No. PBI2715A
  • ONLINE EXCLUSIVE PRICE
    19.76€
    16.75€ / pezzi 1
    Risparmia 3.01€
    15% Sconto
Spedizione stimata: 09-12-2026
Add to basket
Add to basket
Clean Zwiebel™ weights with these anti-static Brushes.
  • Flat synthetic fiber brush
  • Intended for the superficial cleaning of Zwiebel weights
Specifications
Product Type Brush
Length (Metric) 190 mm
Quantity 1 Ea.