missing translation for 'onlineSavingsMsg'
Learn More

LARP6 Antibody, Novus Biologicals™

Product Code. 18614408 Shop All Bio Techne Products
Change view
Click to view available options
Quantità:
25 μL
100 μL
Unit Size:
100µL
25µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantità Unit Size
18614408 25 μL 25µL
18291923 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18614408 Supplier Novus Biologicals Supplier No. NBP25502125ul
 Articolo soggetto a restrizioni
Spedizione stimata: 02-12-2026
Zum Warenkorb hinzufügen
Zum Warenkorb hinzufügen

missing translation for 'validMainframeMsg'

Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Rabbit Polyclonal Antibody

LARP6 Polyclonal specifically detects LARP6 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.
TRUSTED_SUSTAINABILITY

Spezifikation

Antigene LARP6
Applicazioni Immunocytochemistry, Immunofluorescence
Classificazione Polyclonal
Coniugato Unconjugated
Diluizione Immunocytochemistry/Immunofluorescence 1 - 4 μg/mL
Formulazione PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide
Alias gene Acheron, Achn, death-associated LA motif protein, FLJ11196, La ribonucleoprotein domain family member 6, La ribonucleoprotein domain family, member 6, la-related protein 6
Simboli geni LARP6
Specie ospite Rabbit
Immunogeno This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:NMKAVLIGMKPPKKKPAKDKNHDEEPTASIHLNKSLNKRVEELQYMGDESSANSSSDPESNPTSPMAGRRHAATNKLSPSGHQNLFLSPNA
Metodo di purificazione Affinity Purified
Quantità 25 μL
Status giuridico RUO
Primario o secondario Primary
ID gene (immissione) 55323
Test di specificità Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles.
Product Type Antibody
Isotype IgG
Linea di prodotti Novus
Mehr anzeigen Weniger anzeigen

For Research Use Only

Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.