missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ LATEX 400nm Blue Mini Antibody Conjugation Kit

Codice prodotto. 18024624 Sfoglia Tutto Bio Techne Prodotti
Cambia vista
Click to view available options
Quantità:
16 μg
40 μg
Dimensione della confezione:
16µg
pezzi 10
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Quantità Dimensione della confezione
18024624 40 μg pezzi 10
18013854 16 μg 16µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
Codice prodotto. 18024624 Fornitore Novus Biologicals™ N. del fornitore 10000100
 Articolo soggetto a restrizioni
Spedizione stimata: 02-10-2026
Aggiungi al carrello
Aggiungi al carrello

missing translation for 'validMainframeMsg'

Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

The LATEX™ Conjugation Kit allows antibodies or proteins to be conjugated to high quality 400 nm blue latex beads quickly and easily.

The LATEX™ Conjugation Kit allows antibodies or proteins to be conjugated to our high quality 400nm blue latex beads quickly and easily. The latex beads are designed for ease of use and have much improved handling compared with traditional latex. This has been achieved by specially treating the particles. The conjugation is covalent, and requires less antibody than traditional passive and covalent latex conjugates.

Kit Contents:

  • 1 vial of 10x Quencher buffer
  • 1 vial of Resuspension buffer
  • 1 vial of Reaction buffer A
  • 1 vial of Reaction buffer B
  • 1 Midi vial of 400 nm Blue LATEX
TRUSTED_SUSTAINABILITY

Specifications

Bersaglio LATEX™ 400nm Blue Mini
Content And Storage -20°C
Quantità 40 μg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.