Learn More
Novacyt - Microgen Bioproducts Latex Agglutination Test: E.coli
Codice prodotto. 15998012 Sfoglia Tutto Novacyt Group Prodotti
Cambia vista
Click to view available options
Quantità:
50 Tests
Dimensione della confezione:
50 tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Quantità Dimensione della confezione
15998012 50 Tests 50 tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Codice prodotto. 15998012 Fornitore Novacyt - Microgen Bioproducts N. del fornitore M44CE
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Visualizzare prodotti alternativi
Can be used to obtain rapid, presumptive-positive results from selective media for targeted identification/diagnosis.
  • From a range of latex products for the rapid diagnosis of the most common pathogens for foodborne illness.
  • Specific tests available for a variety of infection detections.
  • Rapid (2 minute), simple, sensitive, and specific latex agglutination test for the identification and confirmation of infecting colonies.
  • Can be used to obtain rapid, presumptive-positive results and provide an early indication of possible production process failure.
  • Can be used to screen colonies before full biochemical identification and can replace traditional agglutinating antibody methods.
Specifica
Content And Storage 2°C to 8°C
Analiti rilevabili E. coli
Tipo Latex Agglutination Test
Quantità 50 Tests
Durata del test 2 min.