Learn More
Hach™ LCK Cuvette Tests
Beskrivning
Hach LCK Cuvette Tests are officially approved for legally required consent limits. With the help of Addista standard solutions and round robin test solutions, they provide the assurance that analytical results are always correct. The photometer identifies the cuvette test from its barcode (IBR+) and calculates the result immediately. Cuvettes provide precision and safety using the Dosicap Zip that eliminated contact with reagents.
- Pre-dosed, ready-to-use reagents with no rinsing steps needed
- Precise and reliable measurement values
- Maximum safety for users, thanks to the closed system and low amounts of reagents
- Convenient and error-free dosing of the reagents
- Barcode label for automatic recognition in the photometer
- Differentiation between tests and measuring ranges by means of colour-coding
Beställningsinfo
dimProductType
Specifikationer
Specifikationer
| Analiti | Dissolved Sulphide |
| Certificazione/Conformità | ISO 10530-1991, DIN 38405-D26 |
| Colore | Light blue |
| Formato | Pre-dosed Tubes |
| Da utilizzare con (applicazione) | Water quality testing |
| Da utilizzare con (apparecchiatura) | Photometer (DR3900, DR6000, DR1900, DR2800, DR3800, DR5000), Addista standard solutions |
| Include | Fully equipped box, reagent set |
| N. di test | 25 |
| Range | 0.1 to 2.0mg/L S2- |
| Requisiti di stoccaggio | 15°C to 25°C |
| Visa mer |
Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.