missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LCTL Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Invitrogen PA567189
Dettagli aggiuntivi : Peso : 0.01000kg
Descrizione
Immunogen sequence: SIWDVFTHSGK GKVLGNETAD VACDGYYKVQ EDIILLRELH VNHYRFSLSW Highest antigen sequence identity to the following orthologs - mouse 75%, rat 71%.
This gene encodes a member of family 1 glycosidases. Glycosidases are enzymes that hydrolyze glycosidic bonds and are classified into families based on primary amino acid sequence. Most members of family 1 have two conserved glutamic acid residues, which are required for enzymatic activity. The mouse ortholog of this protein has been characterized and has a domain structure of an N-terminal signal peptide, glycosidase domain, transmembrane domain, and a short cytoplasmic tail. It lacks one of the conserved glutamic acid residues important for catalysis, and its function remains to be determined. Alternative splicing results in multiple transcript variants.Specifica
LCTL | |
Polyclonal | |
Unconjugated | |
LCTL | |
KL lactase phlorizin hydrolase; KLG; klotho gamma; klotho/lactase-phlorizin hydrolase-related protein; KLPH; lactase like; lactase-like; lactase-like protein; LCTL; UNQ3022/PRO9820 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
197021 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q6UWM7 | |
LCTL | |
Recombinant Human LCTL | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
For Research Use Only