Learn More
Taylor Wharton™ LD Series Cryogenic Storage Dewar
Product Code. 11718529
Change view
Click to view available options
Capacity (Metric):
5 L
10 L
50 L
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
11718529 5 L Each
11768519 10 L Each
11798519 50 L Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 11718529 Supplier Taylor Wharton™ Supplier No. LD5
  • ONLINE EXCLUSIVE PRICE
    1075.00€
    997.41€ / pezzi 1
    Risparmia 77.59€
    7% Sconto
 Restricted Item
Spedizione stimata: 16-03-2027
The LD series of cryogenic dewars are used to store and distribute small amounts of liquid nitrogen. They feature extremely low evaporation rates and are convenient to use for various applications.
Specifications
Type Cryogenic Storage Dewar
Capacity (English) 1.3 gal.
Capacity (Metric) 5 L
Height (English) Exterior 17.5 in.
Height (Metric) Exterior 445 mm
Weight (English) 6.8 lb.
Weight (Metric) 3.1 kg