Learn More
3M™ Leather Sweatband HYG4
Product Code. 15739057 Shop All 3M Products
Change view
Click to view available options
Material: Leather
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
15739057 Material: Leather
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15739057 Supplier 3M™ Supplier No. 7000108030
  • ONLINE EXCLUSIVE PRICE
    74.00€
    58.40€ / pezzi 10
    Risparmia 15.60€
    21% Sconto
Spedizione stimata: 09-12-2026
3M™ Leather Sweatband is designed to be used with all 3M™ Hard Hats.
  • Replacement leather sweatband suitable for use with all 3M™ Hard Hats
  • Quick and easy to install
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance CE Approved
Color Beige
For Use With (Equipment) All 3M Safety Helmets
Material Leather
Product Type Leather Sweatband