Learn More
Emma Safety Footwear Leo Safety Shoes
Product Code. 15998821
Change view
Click to view available options
Size:
35
36
37
38
39
48
49
Unit Size:
1 pair
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
15998821 36 1 pair
15988821 35 1 pair
15908831 37 1 pair
15918831 38 1 pair
15928831 39 1 pair
15938831 48 1 pair
15948831 49 1 pair
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Product Code. 15998821 Supplier Emma Safety Footwear Supplier No. MM70054836
  • ONLINE EXCLUSIVE PRICE
    130.84€
    77.02€ / 1 paia
    Risparmia 53.82€
    41% Sconto
Spedizione stimata: 22-12-2026
Includes a steel toe cap, a polyurethane outer toe cap and an easy-twist sole that provides a sticky grip.  Available in a variety of widths.
  • S3-Safety shoe with sole plate for wet work
  • Full grain leather
  • GripForce™ Easy Twist
  • Hydro-Tec™ Sanitized Silver for drier, fresher feet
  • Sanitized
Specifications
Certifications/Compliance EN ISO 20345:2011
Closure Type Laces
Color Black
Design Leo
Liner Material Hydro-Tec™ Sanitized Silver
Material Leather
Size 36
Toe Material Steel