Learn More
3M™ Lexa™ Safety Glasses 15236
Catalog Number 15759787
Click to view available options
Lens Tint: Gray
3M™ Lexa™ Safety Glasses 15236 features both pantoscopic adjustment (angle of the lens frame to the temple) and temple arms with adjustable length (4 fixed positions).
  • Aspheric lens provides expansive coverage and a virtually unobstructed viewing range
  • Frame is fully customizable with different lens angles and temple lengths
  • DX coating provides enhanced anti-scratch and anti-fog protection
  • Polycarbonate lens absorbs 99.9%UV
Lens Coating Scratch-resistant
Lens Tint Clear
Product Type Safety glasses
Colore telaio Black