Learn More
Cytiva LifescienceManual Injection Valve
Product Code. 15239774 Shop All Cytiva Products
Change view
Click to view available options
For Use With (Equipment):
AKTA start Automated Protein Purification System
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
15239774 AKTA start Automated Protein Purification System Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15239774 Supplier Cytiva Supplier No. 29023958
In stock
For use with the AKTA start automated protein purification system. The manual injection valve is a 6-port rotary valve that is manually operated to transfer the preloaded sample onto the column. The sample loop (for the injection of small sample volumes [25μL to 5mL], or a Superloop injection loop for larger sample volumes [10 to 150mL]), can easily be connected to the appropriate ports of the valve.
TRUSTED_SUSTAINABILITY
Specifications
Description 6-port rotary valve that is manually operated to transfer the preloaded sample onto the column
For Use With (Equipment) AKTA start Automated Protein Purification System