missing translation for 'onlineSavingsMsg'
Learn More

Purolite™ Lifetech™ ECR Resins for Enzyme Immobilization, 150 to 300 μm

Lifetech™ ECR resins are robust polymeric resins designed for research and industrial immobilization of enzymes. Available for covalent, ionic, adsorption and affinity immobilization.

Marca:  Purolite™ LS01241-186

Dettagli aggiuntivi : Peso : 0.05000kg

 vedi altre versioni di questo prodotto

Codice prodotto. 17059333

  • 129.00€ / 50g

Spedizione stimata: 11-10-2022
per visualizzare lo stock disponibile
Aggiungi al carrello

Descrizione

Descrizione

  • Enzyme immobilization by adsorption, covalent, IEX and affinity
  • High mechanical stability
  • High porosity
  • Easily dried after immobilization
  • Suitable for batch and column configuration
  • Optimal for use in organic solvent/water free systems
  • Compliant with ResAP(2004) 3
  • Halal, Kosher and TSE/BSE/GMO free
Specifica

Specifica

White to off-white spherical beads
1200 to 1800 Å
150 to 300 μm
Epoxy
ECR8215F
Epoxy Methacrylate
5 to 9
2°C to 8°C
70 to 80%
Wet
50 g
Videos
SDS
Documenti
Certificati
Offerte speciali

Offerte speciali