missing translation for 'onlineSavingsMsg'
Learn More

Purolite™ Lifetech™ Immobilized Enzymes

Lipase immobilized on robust polymeric resins and are supplied in dry form. They can be used for esterifications, amidations, transesterifications, chiral separations. Can be used in water, organic phase, bi-phasic systems and oil phase.

Marca:  Purolite™ LS02040-186

Dettagli aggiuntivi : Peso : 0.01000kg

 vedi altre versioni di questo prodotto

Codice prodotto. 17009403

  • 172.00€ / 10g

Spedizione stimata: 10-10-2022
per visualizzare lo stock disponibile
Aggiungi al carrello

Descrizione

Descrizione

  • Use in solvent-free or organic solvent systems
  • Esterifications (regio- and stereo-selective)
  • Transesterfication
  • Amidation
  • Fats and oils modification
  • Hydrolysis of esters
  • High mechanical stability
  • High enzyme activity
  • Organic solvent stability
  • Available in large volumes
  • Security of supply from Kg to ton scale
  • Resin compliant with ResAP(2004) 3
  • Halal and TSE/BSE/GMO free
Specifica

Specifica

White to slightly yellow spherical beads
300 to 710 μm
Dry
10 g
Lipase from Candida Antarctica B immobilized by adsorption on ECR8806M
2°C to 8°C
<5%
CalB immo™ 8806
Enzyme Activity: >10000 PLU/gdry
Videos
SDS
Documenti
Certificati
Offerte speciali

Offerte speciali