Learn More
Abeba™ Light 31029 Shoes
ESD dissipative shoe includes special respiration absorbing lining for the foot to breathe.
Marca:  Abeba™ 31029/42
Dettagli aggiuntivi : Peso : 1.00000kg
Codice prodotto. 15618387
  • 70.75€ / 1 paio
Spedizione stimata
18-11-2020
Aggiungi al carrello
Descrizione
Descrizione
  • CE EN ISO 20345:2011 S2 SRA
  • Microfiber black
  • Steel toe cap
  • Slip resistance PU outsole sole
  • Replaceable acc Wave insole (article no. 3557)
  • Sanitized treated
  • ESD dissipative according to DIN EN 61340
  • Breathable lining with Silver ions
  • Instep area with elastic band
Specifica
Specifica
CE EN ISO 20345:2011 S2 SRA
Microfiber
Slip-resistant
31029
Black
42
Polyurethane
Steel