Learn More
Abeba™ Light 31055 Shoes
ESD dissipative shoe includes special respiration absorbing lining for the foot to breathe.
Marca:  Abeba™ 31055/46
Dettagli aggiuntivi : Peso : 1.00000kg
Codice prodotto. 15618447
  • 69.65€ / 1 paio
Spedizione stimata
23-11-2020
Aggiungi al carrello
Descrizione
Descrizione
  • CE EN ISO 20345:2011 S1 SRA
  • Leather with Air-Mesh textile inlays
  • Black
  • Steel toe cap
  • Slip resistance PU outsole sole
  • Replaceable acc Wave insole (article no. 3557)
  • Sanitized treated
  • ESD dissipative according to DIN EN 61340
  • Breathable lining with Silver Point
  • Instep area with lacing
  • Reflective material
  • Suitable for heel and foot lift
Specifica
Specifica
CE EN ISO 20345:2011 S1 SRA
Leather
Slip-resistant
31055
Black
46
Polyurethane
Steel