Learn More
Abeba™ Light 31127 Shoes
ESD dissipative shoe includes special respiration absorbing lining for the foot to breathe.
Marca:  Abeba™ 31127/44
Dettagli aggiuntivi : Peso : 0.90000kg
Codice prodotto. 15618597
  • 69.10€ / 1 paio
Spedizione stimata
18-11-2020
Aggiungi al carrello
Descrizione
Descrizione
  • CE EN ISO 20347:2012 O2 FO SRA
  • Microfiber black
  • Slip resistance PU outsole sole
  • Replaceable acc Wave insole (article no. 3559)
  • Sanitized treated
  • ESD dissipative according to DIN EN 61340
  • Breathable lining with Silver ions
  • Instep area with lacing
Specifica
Specifica
CE EN ISO 20347:2012 O2 FO SRA
Microfiber
Slip-resistant
31127
Black
44
Polyurethane