Learn More
Abeba™ Light 31853 Shoes
ESD dissipative shoe includes special respiration absorbing lining for the foot to breathe.
Marca:  Abeba™ 31853/44
Dettagli aggiuntivi : Peso : 1.32700kg
Codice prodotto. 15619327
  • 86.20€ / 1 paio
Spedizione stimata
25-11-2020
Aggiungi al carrello
Descrizione
Descrizione
  • CE EN ISO 20345:2011 S3 SRA
  • Smooth leather
  • Black in ATEX design
  • Steel toe cap
  • Metalfree penetration resistance
  • Slip resistance PU outsole sole
  • Replaceable acc Wave insole (article no. 3557)
  • Sanitized treated
  • ESD dissipative according to DIN EN 61340
  • Breathable lining with Silver Point
  • Instep area with lacing
  • Reflective material
  • Suitable for heel and foot lift
Specifica
Specifica
CE EN ISO 20345:2011 S3 SRA
Leather
Slip-resistant
31853
Black
44
Polyurethane
Steel