Learn More
Novus Biologicals™ Lightning-Link (R) Glucose Oxidase Antibody Labeling Kit
Codice prodotto. 18016304 Sfoglia Tutto Bio Techne Prodotti
Cambia vista
Click to view available options
Quantità:
30 μg
Dimensione della confezione:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Quantità unitSize
18016304 30 μg 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Codice prodotto. 18016304 Fornitore Novus Biologicals™ N. del fornitore 7060030
Please to purchase this item. Need a web account? Register with us today!
For direct labeling of antibodies, proteins, peptides or other biomolecules for use in R&D applications, drug discovery and the development of diagnostic kits
TRUSTED_SUSTAINABILITY
Specifica
Descrizione Antibody Labeling Kit
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Quantità 30 μg
Bersaglio Lightning-Link Glucose Oxidase