missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Lightning-Link (R) PE-Cy5 Antibody Labeling Kit

Codice prodotto. 18790194 Sfoglia Tutto Bio Techne Prodotti
Cambia vista
Click to view available options
Quantità:
180 μg
30 μg
60 μg
600 μg
Dimensione della confezione:
1 set
600µg
60µg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Quantità unitSize
18790194 180 μg 1 set
18026164 600 μg 600µg
18079983 60 μg 60µg
18004994 30 μg 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
Codice prodotto. 18790194 Fornitore Novus Biologicals™ N. del fornitore 7600010
  • ONLINE EXCLUSIVE PRICE
    1060.00€
    1000.65€ / 1 sets
    Risparmia 59.35€
    5% Sconto
 Articolo soggetto a restrizioni
Spedizione stimata: 21-08-2026
Aggiungi al carrello
Aggiungi al carrello
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

Applications: Flow Cytometry

PE/Cy5 is a tandem conjugate. The PE is excited at 488nm and functions as an energy donor for the Cy5. The energy is transferred via FRET (Fluorescence Resonance Energy Transfer) to the Cy5, which emits then the energy received in the form of long wavelength light at 670nm.

The antibody is covalently bonded to the label in a directional and controlled process at near-neutral pH. In view of the large size of the PE-Cy5 label, the amount of antibody used in a labeling reaction with the tandem must always be less than the pack size of LL PE-Cy5, in order that the PE-Cy5 is in a slight molar excess. The best ratio for any new antibody reagent must be determined by experimentation but 50-60ug of IgG antibody for every 100ug of LL PE-Cy5 usually gives optimal results. 50ug of antibody corresponds to an Ab:PE-Cy5 molar ratio of 1:1. The amount of antibody used for labeling should be the same as the pack size of LL-PE-Cy5. For any new antibody, optimization of the ratio of antibody to dye is often worthwhile. If your antibody needs to be concentrated, purified, or cleaned up, consider purchasing an antibody clean up kit.

Order Info

Quantity

TRUSTED_SUSTAINABILITY

Spécification

Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Quantità 180 μg
Coniugato PE/Cy5
Contenuto kit Glass vial(s) of Lightning-Link mix (1 or 3 vials, depending on pack size), 1 vial of LL Modifier reagent, 1 vial of LL Quencher FD reagent
Product Type PE-Cy5 Antibody Labeling Kit
Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.