missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Lightning-Link (R) Streptavidin Antibody Labeling Kit

Codice prodotto. 18064334 Sfoglia Tutto Bio Techne Prodotti
Cambia vista
Click to view available options
Quantità:
100 μg
Dimensione della confezione:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Quantità Dimensione della confezione
18064334 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
Codice prodotto. 18064334 Fornitore Novus Biologicals™ N. del fornitore 7080005
  • ONLINE EXCLUSIVE PRICE
    452.00€
    427.35€ / 100µg
    Risparmia 24.65€
    5% Sconto
 Articolo soggetto a restrizioni
Spedizione stimata: 02-10-2026
Aggiungi al carrello
Aggiungi al carrello

missing translation for 'validMainframeMsg'

Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

Novus Biologicals™ Lightning-Link Streptavidin Antibody Labeling Kit enables the direct conjugation of streptavidin to any biomolecule with an available amine group.

  • Enable the direct labeling of antibodies, proteins, peptides or other biomolecules for use in research and development applications, drug discovery and the development of diagnostic kits.
  • Researcher simply pipettes their antibody or other biomolecule into the vial of Lightning-Link Streptavidin and incubates for 3 hours.
  • Streptavidin is a 53kDa protein purified from Streptomyces avidinii, which has widespread applications due to its very high affinity to the vitamin biotin. Streptavidin is a tetrameric molecule, composed of 4 13kDa monomers, each of which can bind a molecule of biotin.
  • The kit is unique in the sense that it is the only DIY streptavidin labeling kit available on the market. Like our biotinylation kits, Lightning-Link Streptavidin has been optimised to create high quality conjugates and minimise assay background. There is also no need for a wash or desalting step. The way in which the kits are optimised means the antibody or protein of interest will be labeled with up to 3 streptavidin molecules each, maximising the opportunity for biotin to bind and create a biotin streptavidin complex.
TRUSTED_SUSTAINABILITY

Specifica

Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Quantità 100 μg
Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.