Learn More
Novus Biologicals™ Lightning-Link (R) Streptavidin Antibody Labeling Kit
Codice prodotto. 18785213 Sfoglia Tutto Bio Techne Prodotti
Cambia vista
Click to view available options
Quantità:
1 mg
30 μg
300 μg
Dimensione della confezione:
1 set
1mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Quantità unitSize
18785213 30 μg 1 set
18736483 300 μg 1 set
18745363 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Codice prodotto. 18785213 Fornitore Novus Biologicals™ N. del fornitore 7080030
  • ONLINE EXCLUSIVE PRICE
    625.00€
    591.15€ / 1 sets
    Risparmia 33.85€
    5% Sconto
 Articolo soggetto a restrizioni
Spedizione stimata: 03-08-2026
Aggiungi al carrello
Aggiungi al carrello
Antibody labeling kit
Streptavidin is a 53kDa protein purified from Streptomyces avidinii, which has widespread applications due to its very high affinity to the vitamin biotin. Streptavidin is a tetrameric molecule, composed of 4 13kDa monomers, each of which can bind a molecule of biotin. The Lightning-Link Streptavidin kit is unique in the sense that it is the only DIY streptavidin labeling kit available on the market. Like our biotinylation kits, Lightning-Link Streptavidin has been optimised to create high quality conjugates and minimise assay background. There is also no need for a wash or desalting step. The way in which the kits are optimised means the antibody or protein of interest will be labeled with up to 3 streptavidin molecules each, maximizing the opportunity for biotin to bind and create a biotin streptavidin complex. If your antibody needs to be concentrated, purified, or cleaned up, consider purchasing an antibody clean up kit.
Order Info
Quantity
TRUSTED_SUSTAINABILITY
Specifica
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Quantità 30 μg
Contenuto kit Glass vial(s) of Lightning-Link mix (1 or 3 vials, depending on pack size), 1 vial of LL-AP Modifier reagent, 1 vial of LL-AP Quencher reagent
Product Type Streptavidin Antibody Labeling Kit