Learn More
α-Lipoic Acid, MedChemExpress
Product Code. 16402386
Encompass_Preferred
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16402386 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 16402386 Supplier Medchem Express Supplier No. HYN0492
 Restricted Item
Spedizione stimata: 12-01-2027
MedChemExpress α-Lipoic Acid is an antioxidant, which is an essential cofactor of mitochondrial enzyme complexes. α-Lipoic Acid inhibits NF-κB-dependent HIV-1 LTR activation. α-Lipoic Acid induces endoplasmic reticulum (ER) stress-mediated apoptosis in hepatoma cells.
α-Lipoic Acid is an antioxidant, which is an essential cofactor of mitochondrial enzyme complexes. α-Lipoic Acid inhibits NF-κB-dependent HIV-1 LTR activation. α-Lipoic Acid induces endoplasmic reticulum (ER) stress-mediated apoptosis in hepatoma cells.
Specifications
Chemical Name or Material α-Lipoic Acid
CAS 1077-28-7
Color Yellow
Purity Grade Notes Research
Molecular Formula C8H14O2S2
Quantity 500 mg
Synonym Thioctic acid (±)-α-Lipoic acid DL-α-Lipoic acid
Solubility Information DMSO : ≥ 100 mg/mL (484.66 mM) ∣H2O : 0.1 mg/mL (0.48 mM; Need ultrasonic)
SMILES O=C(O)CCCCC1SSCC1
Molecular Weight (g/mol) 206.33
Formula Weight 206.33
Percent Purity 97.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less
Research purposes only