Learn More
Thermo Scientific™ LN2 Supply Tanks
Transport cryogenic industrial gases with these LN2 Supply tanks, which are designed to be both durable and user-friendly.
Specifica
Tipo LN2 Supply Tank
Visualizza altre specifiche
Descrizione
Descrizione
Transport cryogenic industrial gases with Thermo Scientific™ LN2 Supply tanks, which are designed to be both durable and user-friendly.
For long-term storage of samples in cryogenic freezers, or controlling temperature during freezing procedures utilizing controlled-rate freezing equipment.
  • Rated at 22psi (151.7kPa)
  • Evaporate 2% of volume per day
  • Warranty varies by country, please contact us for details
Specifica
Specifica
LN2 Supply Tank