Learn More
Astell Scientific™ Load Support Plate
Support shelf to expand autoclave capacity. Astell Scientific™ Load Support Plate accommodates additional Duran bottles without the need for baskets.
Marca:  Astell Scientific™ AAN074
Dettagli aggiuntivi : Peso : 1.00000kg
Codice prodotto. 12795355
  • 202.00€ / Pezzo
Spedizione stimata
17-03-2021
Aggiungi al carrello
Descrizione
Descrizione
  • Expands autoclave loading capacity.
  • Supports additional Duran bottles without the need for baskets.
  • Stainless steel construction.
Specifica
Specifica
AMA250, AMA260, AMA270 autoclaves
Load support plate
Stainless steel
1Each