Learn More
Thermo Scientific™ Locator™ Plus Rack and Box Systems
Codice prodotto. 10307713 Sfoglia Tutto Thermo Scientific Prodotti
Cambia vista
Click to view available options
Capacità (sistema metrico):
71 L
121 l
184 L
Modello:
Locator 4 Plus
Locator 6 Plus
Locator 8 Plus
Locator Jr. Plus
Include:
Con monitor di livello
With Level Monitor
Without Level Monitor
Capacità del criotubo:
2000
4000
6000
Dimensione della confezione:
Pezzo
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Capacità (sistema metrico) Modello Include Capacità del criotubo Dimensione della confezione
10307713 121 l Locator 4 Plus Con monitor di livello 4000 Pezzo
11776359 71 L Locator Jr. Plus With Level Monitor 2000 Pezzo
11716369 184 L Locator 6 Plus With Level Monitor 6000 Pezzo
11736369 121 l Locator 8 Plus With Level Monitor 2000 Pezzo
10063553 71 L Locator Jr. Plus Without Level Monitor 2000 Pezzo
10533874 121 l Locator 4 Plus Without Level Monitor 4000 Pezzo
10398533 121 l Locator 8 Plus Without Level Monitor 2000 Pezzo
10028132 184 L Locator 6 Plus Without Level Monitor 6000 Pezzo
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Codice prodotto. 10307713 Fornitore Thermo Scientific™ N. del fornitore CY509108
  • ONLINE EXCLUSIVE PRICE
    8885.00€
    8580.00€ / pezzi 1
    Risparmia 305.00€
    3% Sconto
Visualizza tutte le promozioni
 Articolo soggetto a restrizioni
Spedizione stimata: 09-12-2026
Economically store up to 6,000 samples in the Locator 6 rack and box system that makes sample retrieval simple and consumes minimum liquid nitrogen.
Economically store up to 6,000 samples in the Thermo Scientific™ Locator™ Plus Rack and Box System that makes sample retrieval simple and consumes minimum liquid nitrogen. This indexed system is available with or without ultrasonic level monitor.
Features:
  • Compact design to minimize space requirements while maximizing storage capacity
  • Suited for both manual and computerized inventory record-keeping methods
  • Outstanding temperature uniformity: samples are stored below -180°C, even when less than 2' (5cm) of liquid nitrogen remains in the vessel
  • Audible alarm that sounds when nitrogen level falls below safe range; dry remote alarm contact for remote monitoring
  • Advanced vacuum insulation to minimize liquid nitrogen evaporation and reduces operating costs
  • Secure locking to prevent unauthorized entry
Locator 4 Plus and Locator 8 Plus:
  • With level monitor
  • Outside Dia.: 55.8cm; Height 100.3cm
Locator Jr. Plus:
  • Without Level Monitor
  • Outside Dia.: 55.9cm; Height: 67.9cm
Locator 6 Plus:
  • Without Level Monitor
  • Outside Dia.: 66cm; Height :100.3cm
Note:
Stated vial capacity assumes 1.2/2mL Thermo Scientific™ Nalgene™ Cryogenic Vials
Includes:
Indexed stainless-steel racks designed for use with 2' (5cm) Nalgene™ Cryogenic Boxes (larger racks available for storage of 5.0mL ampules in Locator Jr. Plus and Locator 4 Plus). Available with or without Liquid Nitrogen Level Monitor (optional, factory-installed monitor can also be purchased separately).
Requires:
Thermo Scientific Nalgene Cryogenic Boxes (order separately)
Compatible with:
  • Boxes, Nalgene, 2' for 1.0 and 1.5mL vials, 100 vials per box, case of 10 (CS509X24)
  • Boxes, Nalgene, 2' for 2mL vials, 81 vials per box, case of 40 (CS509X4); case of 20 (CS509X5)
  • Boxes, Nalgene, 4' for 5mL vials, 81 vials per box, each (CS509X10)
  • Thermo Scientific™ CryoClaw™ (AY509X18)
  • Cups, Suction, 2 replacements for CryoClaw (AY509X19)
  • Logbook (LT509X5)
  • Level Monitor, customer-installed: CN509X3-70 (120V, Jr.); CN509X6-70 (120V, 4); CN509X10-70 (120V, 6); CN509X9-70 (120V, 8); CN509X1-70 (220V, Jr.); CN509X4-70 (220V, 4); CN509X11-70 (220V, 6); CN509X7-70 (220V, 8)
  • Rack, for 5.0mL vials (does not include boxes) (HR509X19A)
  • Vials, 1mL, case of 500 (AY509X32)
  • Vials, Cryogenic, 1.2mL, case of 500, 1.2 ml (AY509X13)
  • Vials, 1.5mL, case of 500 (AY509X33)
  • Vials, Cryogenic, 2.0mL, case of 500, 2.0 ml (AY509X12)
TRUSTED_SUSTAINABILITY
Specifica
Tipo Manual Fill LN2 Dewar
Capacità (sistema imperiale) 4.273 cu. ft.
Capacità (sistema metrico) 121 l
Gamma di temperatura Below -180°C
Isolamento Vacuum
Certificazione/Conformità CE, CSA
Include Con monitor di livello
Modello Locator 4 Plus
Autonomia fase statica 122 giorni
Altezza (sistema imperiale) esterno 37.5 in.
Altezza (sistema metrico) esterno 95.3 cm
Diametro (sistema imperiale) 22 in.
Diametro (sistema imperiale) interno collo 8.5 in.
Diametro (sistema metrico) 55.9 cm
Diameter (Metric) Inside Neck 21.5 cm
Opzioni di monitoraggio Ultrasonic Level
Tensione 220 V
Requisiti elettrici 220 V 50 Hz
Frequenza 50 Hz
Utilizzo di energia 0.60 l
Capacità del criotubo 4000
Peso di spedizione (sistema metrico) 53.5 kg
Capacità Samples (4000)
Capacità rack 4