missing translation for 'onlineSavingsMsg'
Learn More

Uvex™ Long John 7483-Black

Catalog Number 17286051
Click to view available options
Size:
3X-Large/4X-Large
5X-Large/6X-Large
Medium/Large
Small
X-Large/2X-Large
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

Well thought-out functionality, sporty design, exciting color contrasts, unusual material combinations and a cut that fits like a glove.

  • Long trousers merino men uvex suXXeed
  • Color: Black
  • Material: 60% polypropylene, 28% wool Merino, 8% PA, 4% EL
  • Seamless underwear, without annoying seams, moisture-regulating, with optimal fit for maximum comfort
  • Soft cuffs, merino wool for best insulation, odor neutralizing
  • Size: S, M/L, XL/XXL, 3XL/4XL, 5XL/6XL
Size 3X-Large/4X-Large
Colore Black
Spessore (sistema metrico) 250 g/m²
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'