Learn More
Lornoxicam, TRC
Product Code. 18000620
Change view
Click to view available options
Quantity:
10 mg
50 mg
250 mg
Unit Size:
10mg
250mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18000620 50 mg 50mg
18093410 10 mg 10mg
18032620 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 18000620 Supplier Toronto Research Chemicals Supplier No. TRCL47000050MG
 Restricted Item
Spedizione stimata: 11-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 70374-39-9
Molecular Formula C13 H10 Cl N3 O4 S2
Molecular Weight (g/mol) 371.82
Synonym 6-Chloro-4-hydroxy-2-methyl-N-2-pyridinyl-2H-thieno[2,3-e]-1,2-thiazine-3-carboxamide 1,1-dioxide, Chlortenoxicam, Fulactive, Lofecam, Lorcam, Lornoxicam, Ro 13-9297, TS 110, Telos, Xefo, Xefocam
IUPAC Name 6-chloro-4-hydroxy-2-methyl-1,1-dioxo-N-pyridin-2-ylthieno[2,3-e]thiazine-3-carboxamide
SMILES CN1C(=C(O)c2sc(Cl)cc2S1(=O)=O)C(=O)Nc3ccccn3
Specifications
Quantity 50 mg
InChI Formula InChI=1S/C13H10ClN3O4S2/c1-17-10(13(19)16-9-4-2-3-5-15-9)11(18)12-7(23(17,20)21)6-8(14)22-12/h2-6,18H,1H3,(H,15,16,19)
Formula Weight 370.9801
Percent Purity >95%
Chemical Name or Material Lornoxicam
Grade HPLC