Learn More
Lovibond™ Comparatore 705
Versatile and highly precise visual colorimetric water analysis system for rapid and accurate water analysis and color measurement
Marca: Lovibond™ 142000
Dettagli aggiuntivi : Peso : 0.26000kg
Descrizione
Versatile and highly precise visual colorimetric water analysis system for rapid and accurate water analysis and color measurement
Ideal for testing water, wastewater, boiler and cooling water, process fluids, swimming pools, spas and more in the lab, plant, field or classroom. Handheld instrument is easy to use and provides precise, reproducible results.
- Handheld and completely portable
- Can be used for a wide variety of color measurement applications
- Integrated magnifying prism brings standard and test sample colors side by side and into one image, helping to make an exact color match
- Adjustable sample cell chamber can accommodate cells from 10 to 40mm path lengths
- Simple one-button press operation
- Compensation for colored or turbid samples
- Consistency guaranteed with permanent colored glass standards
- Color discs are fade-free, chemical-resistant and scratch-proof; comply with ISO 7392/2
- Uses 13mm, 10mL tubes (600-T13P)
- A Nesslerizer System is required for specified tests in order to obtain accurate results.
Specifica
ISO 7393-2 | |
1 involucro per comparatore 705. Le celle campione sono vendute separatamente. |