Learn More
Lovibond™ Comparatore 705
Codice prodotto. 11763187
Cambia vista
Click to view available options
Certificazione/Conformità:
ISO 7393-2
Dimensione della confezione:
Pezzo
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Certificazione/Conformità Dimensione della confezione
11763187 ISO 7393-2 Pezzo
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Codice prodotto. 11763187 Fornitore Lovibond™ N. del fornitore 142000
  • ONLINE EXCLUSIVE PRICE
    200.00€
    169.40€ / pezzi 1
    Risparmia 30.60€
    15% Sconto
Visualizza tutte le promozioni
Spedizione stimata: 08-02-2027
Versatile and highly precise visual colorimetric water analysis system for rapid and accurate water analysis and color measurement
Ideal for testing water, wastewater, boiler and cooling water, process fluids, swimming pools, spas and more in the lab, plant, field or classroom. Handheld instrument is easy to use and provides precise, reproducible results.
  • Handheld and completely portable
  • Can be used for a wide variety of color measurement applications
  • Integrated magnifying prism brings standard and test sample colors side by side and into one image, helping to make an exact color match
  • Adjustable sample cell chamber can accommodate cells from 10 to 40mm path lengths
  • Simple one-button press operation
  • Compensation for colored or turbid samples
  • Consistency guaranteed with permanent colored glass standards
  • Color discs are fade-free, chemical-resistant and scratch-proof; comply with ISO 7392/2
  • Uses 13mm, 10mL tubes (600-T13P)
  • A Nesslerizer System is required for specified tests in order to obtain accurate results. 
Note: Tubes and Color Discs are not included; sold separately.  (For tubes, see 600-T13P.)  
Specifica
Certificazione/Conformità ISO 7393-2
Include 1 involucro per comparatore 705. Le celle campione sono vendute separatamente.