Learn More
Lovibond™ PFXi-995 Colorimeter: Oils, Chemicals and Derivatives
Codice prodotto. 11953709
Cambia vista
Click to view available options
Tipo:
Colorimeter
Dimensione della confezione:
Pezzo
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Tipo unitSize
11953709 Colorimeter Pezzo
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Codice prodotto. 11953709 Fornitore Lovibond™ N. del fornitore 1379951
High-precision, automatic spectrophotometer for objective color measurement
Accurate, highly efficient spectrophotometer for optically clear samples meets the demand for consistent and reliable color data for a wide range of samples. The long path-length cell chamber also allows the measurement of extremely unsaturated, light samples and also color scales which are defined with long path lengths including the Lovibond RYBN and AOCS Tintometer™ color. The instrument removes all subjectivity involved in color grading, supplying unbiased readings that are unaffected by operator or environment.
  • Color Scales: Lovibond RYBN, AOCS-Tintometer, Gardner, Pt-Co/Hazen/APHA, FAC, Chlorophyll A and B, beta Carotene, Iodine, Hess-Ives Color Units, Klett (blue filter KS-42), Kreis Value (rancidity), CIE Values, Spectral Data
  • Automatic and nonsubjective color measurement
  • Consistent data compliant with international standards
  • Rugged, chemically resistant, steel construction
  • Easy maintenance with removable sample chamber
  • Ability to handle hot samples with high melting points
  • Extensive and flexible choice of color scales
  • Remote upgrade capability
  • Calculation and description of off-hue status
  • Supplied with color control software for data analysis
  • Conformance standard, cells and spare bulb included
  • Output conforming to GLP including date, time, sample and user ID
  • Accommodates a wide range of sample cells and tubes
  • Wide range of accessories is available (e.g. thermal printer)
  • 6 in. (153mm) sample chamber guarantees compliance with AOCS, ISO and ASTM standards
  • Available with integrated RCMSi technology for remote calibration
  • Optional integrated heater unit, up to 95°C
Compliance
Complies with AOCS, ISO and ASTM standards.
Specifica
Calibrazione Yes
Certificazione/Conformità CE, RoHS, WEEE
Collegamenti USB, LAN, RS232
Memorizzazione dei dati 100,000+ Measurements
Display Backlit LCD
Dimensioni (L x P x A) 7.71 x 20.27 x 6.69 in. (19.6 x 51.5 x 17 cm)
Lingue English, French, German, Italian, Spanish, Portuguese, Russian, Chinese, Japanese
Sorgente luminosa Tungsten Halogen
Larghezza di banda spettrale 10 nm
Tipo Colorimeter
Tensione 90/240 V
Wavelength Range 420 to 710 nm
Da utilizzare con (applicazione) Transparent samples
Altezza (sistema imperiale) 6.69 in.
Altezza (sistema metrico) 17 cm
Include Windows™ compatible software, spare lamp, instructions, RCMSi starter pack and Lovibond™ optical Glass cells
Lunghezza (sistema imperiale) 7.71 in.
Lunghezza (sistema metrico) 19.6 cm
Materiale Textured Fabricated Steel
Lunghezza del percorso 1 x 10, 1 x 100 mm
Peso (sistema imperiale) 16.9 lb.
Peso (sistema metrico) 7.7 kg
Larghezza (sistema imperiale) 20.27 in.
Larghezza (sistema metrico) 51.5 cm