Learn More
Brand™ Low-Form Polypropylene Beakers with Embossed Scale
Catalog Number p-8015492
Click to view available options
Capacità (sistema metrico):
50 mL
100 mL
400 mL
2000 mL
3000 mL
Transparent
  • With beaded rim and spout
  • Autoclavable
Capacità (sistema metrico) 50 mL
Materiale Polypropylene
Incrementi 10mL
Graduated Yes