Learn More
Loxoprofen, TRC
Product Code. 17962125
Change view
Click to view available options
Quantity:
25 mg
100 mg
1 g
Unit Size:
100mg
1g
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17962125 1 g 1g
17901175 25 mg 25mg
17952085 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 17962125 Supplier Toronto Research Chemicals Supplier No. TRCL4729051G
 Restricted Item
Spedizione stimata: 11-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 68767-14-6
Molecular Formula C15 H18 O3
Molecular Weight (g/mol) 246.3
Synonym Benzeneacetic acid, α-methyl-4-[(2-oxocyclopentyl)methyl]-, α-Methyl-4-[(2-oxocyclopentyl)methyl]benzeneacetic acid, 2-[4-[(2-Oxocyclopentan-1-yl)methyl]phenyl]propionic acid, 2-[4-[(2-Oxocyclopentyl)methyl]phenyl]propanoic acid, 2-[4-[(2-Oxocyclopentyl)methyl]phenyl]propionic acid, Loxoprofen
IUPAC Name 2-[4-[(2-oxocyclopentyl)methyl]phenyl]propanoic acid
SMILES CC(C(=O)O)c1ccc(CC2CCCC2=O)cc1
Specifications
Quantity 1 g
InChI Formula InChI=1S/C15H18O3/c1-10(15(17)18)12-7-5-11(6-8-12)9-13-3-2-4-14(13)16/h5-8,10,13H,2-4,9H2,1H3,(H,17,18)
Formula Weight 246.1256
Percent Purity >95%
Chemical Name or Material Loxoprofen
Grade HPLC