Learn More
Merck LpDNPH Ozone Scrubber S10L (Reversible Cartridge), Potassium Iodide (KI)
Product Code. 17709817 Shop All Merck Products
Change view
Click to view available options
Quantity:
10/Pk.
Unit Size:
Pack of 10
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17709817 10/Pk. Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17709817 Supplier Merck Supplier No. 505285
 Restricted Item
This item requires a price update prior to being ordered.
Request a Price for this item here.
missing translation for 'validMainframeMsg'
This item is not returnable. View return policy
Ozone scrubbers are recommended for sampling carbonyls in high humidity environments.
LpDNPH cartridges contain 1.5 g of high purity potassium iodide (KI) and are available in two cartridge styles. KI traps the ozone which causes a negative formaldehyde interference in DNPH coated devices. The reversible cartridge offers a slip luer end-fitting, and the rezorian cartridge offers a luer lock end-fitting, to enable you to connect your cartridge of choice directly to the inlet of any DNPH cartridge with a like end-fitting. Testing (200 ppb ozone, 50 % RH, 25C) indicates the ozone scrubber to have an ozone capacity of 100,000 ppb/hr.
TRUSTED_SUSTAINABILITY
Specifications
Description Potassium Iodide (KI), 1.5 g
Quantity 10/Pk.
Product Title
Select an issue
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.