Learn More
LPHN3 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Thermo Scientific PA553381
Dettagli aggiuntivi : Peso : 0.02000kg
Descrizione
Immunogen sequence: LCLAPDGIWD PQGPDLSNCS SPWVNHITQK LKSGETAANI ARELAEQTRN HLNAGDITYS VRAMDQLVGL LDVQLRNLTP GGKDSAARSL N Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 100%.
LPHN3 Polyclonal antibody specifically detects LPHN3 in Human samples. It is validated for Immunohistochemistry.Specifica
LPHN3 | |
Polyclonal | |
Unconjugated | |
ADGRL3 | |
5430402I23Rik; ADGRL3; Adhesion G protein-coupled receptor L3; calcium-independent alpha-latrotoxin receptor 3; calcium-independent alpha-latrotoxin receptor homolog 3; Cirl3; CIRL-3; Cl3; D130075K09Rik; Gm1379; Kiaa0768; latrophilin 3; latrophilin homolog 3 (cow); latrophilin-3; LEC3; lectomedin 3; lectomedin-3; Lph3; LPHN3; mKIAA0768 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
23284 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
Lot-Specific | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9HAR2 | |
ADGRL3 | |
Recombinant protein corresponding to Human LPHN3 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only