Learn More
LRRC23 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Thermo Scientific PA558095
Dettagli aggiuntivi : Peso : 0.02000kg
Descrizione
Immunogen sequence: QNMLKKVEGL EDLSNLTTLH LRDNQIDTLS GFSREMKSLQ YLNLRGNMVA NLGELAKLRD LPKLRALVLL DNPCTDETSY R Highest antigen sequence identity to the following orthologs: Mouse - 85%, Rat - 86%.
LRRC23 Polyclonal antibody specifically detects LRRC23 in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.Specifica
LRRC23 | |
Polyclonal | |
Unconjugated | |
LRRC23 | |
4921537K05Rik; B7; leucine rich protein, B7; leucine rich repeat containing 23; leucine-rich protein B7; Leucine-rich repeat-containing protein 23; LRPB7; LRRC23; neuron-specific enolase | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
10233 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry, Western Blot | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q53EV4 | |
LRRC23 | |
Recombinant protein corresponding to Human LRRC23 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only