Learn More
Corning™ LSE™ Fixed-Angle Rotors
Product Code. 11367681 Shop All Corning Products
Change view
Click to view available options
Capacity:
18 x 1.5 mL
6 x 50 mL
12 x 15 mL
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity Unit Size
11367681 6 x 50 mL Each
11397681 18 x 1.5 mL Each
11377681 12 x 15 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 11367681 Supplier Corning™ Supplier No. 480136
  • ONLINE EXCLUSIVE PRICE
    1069.63€
    959.57€ / pezzi 1
    Risparmia 110.06€
    10% Sconto
 Restricted Item
Spedizione stimata: 09-04-2027
Rotors with different tube size and capacity configurations
This is an optional rotor for 6 x 50 mL tubes, for use with Corning™ LSETM Compact Centrifuge.
TRUSTED_SUSTAINABILITY
Specifications
Type Rotor
Capacity 6 x 50 mL
Max. Speed 6,000 rpm
For Use With (Equipment) Corning™ LSE™ Compact Centrifuge
Angle Fixed