Learn More
Chem Lab™ Luff-Schoorl Reagent
Product Code. 15137542
Change view
Click to view available options
Quantity:
1 L
Unit Size:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15137542 1 L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15137542 Supplier Chem Lab™ Supplier No. CL02.1207.1000
  • ONLINE EXCLUSIVE PRICE
    61.00€
    28.50€ / 1L
    Risparmia 32.50€
    53% Sconto
 Restricted Item
Spedizione stimata: 30-11-2026
Specifications
Quantity 1 L
compliance-icons compliance-icons
Product Identifier
  • Luff-Schoorl reagent
Signal Word
  • Warning
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
  • H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: Sodium carbonate