Learn More
LUZP1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Thermo Scientific PA555883
Dettagli aggiuntivi : Peso : 0.02000kg
Descrizione
Immunogen sequence: RERHTSTSNI QVGLAELTSV SNHVSSPFEL SIHKHDITLQ LAEAERMADG PLKNRPETVV SRSSIIIKPS DPVERNSHAP PAETIRWKSH Highest antigen sequence identity to the following orthologs: Mouse - 81%, Rat - 86%.
LUZP1 Polyclonal antibody specifically detects LUZP1 in Human samples. It is validated for Immunohistochemistry.Specifica
LUZP1 | |
Polyclonal | |
Unconjugated | |
LUZP1 | |
2700072H04Rik; AI266952; Leucine zipper motif-containing protein; leucine zipper protein 1; Luzp; LUZP1; mFLJ00226; RP1-184J9.3 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
7798 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q86V48 | |
LUZP1 | |
Recombinant protein corresponding to Human LUZP1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only