Learn More
LysoPC(14:0/0:0), MedChemExpress
Product Code. 16447477
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16447477 5 mg 5mg
16404758 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 16447477 Supplier Medchem Express Supplier No. HY113123
 Restricted Item
Spedizione stimata: 04-12-2026
MedChemExpress LysoPC(14:0/0:0) is a lysophospholipid (LyP). It is a monoglycerophospholipid in which a phosphorylcholine moiety occupies a glycerol substitution site. LysoPC(14:0/0:0) has potent antispasmodic effect.
LysoPC(14:0/0:0) is a lysophospholipid (LyP). It is a monoglycerophospholipid in which a phosphorylcholine moiety occupies a glycerol substitution site. LysoPC(14:0/0:0) has potent antispasmodic effect.
Specifications
Chemical Name or Material LysoPC(14:0/0:0)
CAS 20559-16-4
Color White
Purity Grade Notes Research
Molecular Formula C22H46NO7P
Quantity 5 mg
Solubility Information Ethanol : 50 mg/mL (106.93 mM; Need ultrasonic)
SMILES CCCCCCCCCCCCCC(OC[C@@H](O)COP(OCC[N+](C)(C)C)([O-])=O)=O
Molecular Weight (g/mol) 467.58
Formula Weight 467.58
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Metabolism-sugar/lipid metabolism
Show More Show Less
Research purposes only