missing translation for 'onlineSavingsMsg'
Learn More

m-Tolyl acetate, 97%, Thermo Scientific Chemicals

Catalog Number p-7046499
Click to view available options
Quantità:
25 g
100 g
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
122-46-3
C9H10O2
150.18
MFCD00041910
OTGAHJPFNKQGAE-UHFFFAOYSA-N
m-tolyl acetate, m-cresyl acetate, cresatin, m-acetoxytoluene, m-cresol acetate, acetic acid m-tolyl ester, cresatin-sulzberger, kresatin, acetic acid, 3-methylphenyl ester, m-methylphenyl acetate
67406
(3-methylphenyl) acetate
CC(=O)OC1=CC=CC(C)=C1
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
122-46-3
C9H10O2
150.18
MFCD00041910
OTGAHJPFNKQGAE-UHFFFAOYSA-N
m-tolyl acetate, m-cresyl acetate, cresatin, m-acetoxytoluene, m-cresol acetate, acetic acid m-tolyl ester, cresatin-sulzberger, kresatin, acetic acid, 3-methylphenyl ester, m-methylphenyl acetate
67406
(3-methylphenyl) acetate
CC(=O)OC1=CC=CC(C)=C1

m-Tolyl acetate as a pharmaceutical intermediate and antiseptic.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
m-Tolyl acetate as a pharmaceutical intermediate and antiseptic.

Solubility
Not miscible or difficult to mix with water.

Notes
Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY
Materiale o nome chimico m-Tolyl acetate
Densità 1.05
Punti di ebollizione 100°C (14 mmHg)
Punto d'infiammabilità 95°C (203°F)
Indice di rifrazione 1.502
Quantità 25 g
Beilstein 1862796
Indice di Merck 14,2583
Informazioni di solubilità Not miscible or difficult to mix with water.
Peso formulazione 150.18
Percent Purity 97%
Vedi altri risultati Mostra meno risultati

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'