missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ m-Tolylacetic acid, 99%

Marca:  Alfa Aesar™ A17743.30

Dettagli aggiuntivi : CAS : 621-36-3 Peso : 0.25000kg

Quantità 250g

 vedi altre versioni di questo prodotto

Codice prodotto. 11403348

  • 486.00€ / 250g


Spedizione stimata
27-04-2021
Aggiungi al carrello

Descrizione

Descrizione

Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Soluble in water.

Notes
Store in a tightly closed container in a cool, dry, well-ventilated area. Store away from strong oxidizing agents.
Specifica

Identificatori chimici

621-36-3
150.177
GJMPSRSMBJLKKB-UHFFFAOYSA-N
12121
CC1=CC(=CC=C1)CC(=O)O
C9H10O2
MFCD00004340
3-methylphenylacetic acid, m-tolylacetic acid, 2-m-tolyl acetic acid, 2-3-methylphenyl acetic acid, 3-tolylacetic acid, m-methylphenylacetic acid, 3-methylphenyl acetic acid, m-tolylaceticacid, benzeneacetic acid, 3-methyl, methyll 3-methylphenylacetate
2-(3-methylphenyl)acetic acid

Specifica

m-Tolylacetic acid
250g
C9H10O2
2043549
Soluble in water.
CC1=CC(=CC=C1)CC(=O)O
150.177
150.18
62°C to 66°C
621-36-3
MFCD00004340
3-methylphenylacetic acid, m-tolylacetic acid, 2-m-tolyl acetic acid, 2-3-methylphenyl acetic acid, 3-tolylacetic acid, m-methylphenylacetic acid, 3-methylphenyl acetic acid, m-tolylaceticacid, benzeneacetic acid, 3-methyl, methyll 3-methylphenylacetate
GJMPSRSMBJLKKB-UHFFFAOYSA-N
2-(3-methylphenyl)acetic acid
12121
99%
Videos
SDS Documenti