Descrizione
To ensure incredibly fast and effective sample heating the MA35 is equipped with two powerful metal rod dark infrared radiators. Easy to understand icons guide you in three steps from taring the sample pan to starting the measurement. All important operating parameters can be accessed and changed in seconds.
- 40°C to 160°C temperature range, adjustable in increments of 1°C
- Fully proven, reliable and effective dark infrared radiator for sample heating
- Easy to operate by clear, easy to follow symbol guidance
- Fully automatic end point detection or 0.1 to 99 minutes timer shut off mode
Specifica
Specifica
| Certificazione/Conformità | Regolamento FDA/HACCP |
| Display | Digitale |
| Modello | MA35M-000230V1 |
| Requisiti di alimentazione | 230 V |
| Sorgente di radiazioni | Metal Rod Heater |