Learn More
Brady Magicard™ CR80 PVC Card with HoloPatch
Product Code. 30067370 Shop All Brady Products
Change view
Click to view available options
Color:
White
Unit Size:
Pack of 500
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Unit Size
30067370 White Pack of 500
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30067370 Supplier Brady Supplier No. 322002
  • ONLINE EXCLUSIVE PRICE
    134.00€
    120.23€ / pezzi 500
    Risparmia 13.77€
    10% Sconto
Spedizione stimata: 01-02-2027
Plain white PVC Card (CR80) with HoloPatch™ for extra visual security.
  • Pre-approved for best quality printing on Magicard Printers.
  • HoloPatch™ for extra visual security.
  • Pack of 500 cards.
Specifications
Quantity 500 Pk.
Product Type Badge
Alternate Part Number M9006-796
Color White
Material PVC
Message Blank
For Use With (Application) Healthcare and Hospital labelling;General identification;Facility identification;Security identification
Height (English) 3.4 in.
Height (Metric) 86 mm
Thickness 0.762 mm
Width (English) 2.1 in.
Width (Metric) 54 mm
Label Type Cards for Printers
For Use With (Equipment) Magicard 300, Magicard 600, Pronto 100, Prima 8