Learn More
Magnesium carbonate hydroxide pentahydrate, light, 98%
Product Code. 15477515
Change view
Click to view available options
Quantità:
250 g
1000 g
5000 g
Unit Size:
1000g
250g
5000g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantità Unit Size
15477515 1000 g 1000g
15497515 250 g 250g
15487515 5000 g 5000g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 15477515 Supplier Thermo Scientific Alfa Aesar Supplier No. A18070.0B
 Articolo soggetto a restrizioni
In stock
Add to basket
CAS: 56378-72-4 | C4H12Mg5O19 | 485.65 g/mol
Magnesium carbonate hydroxide pentahydrate is used as pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Magnesium carbonate hydroxide pentahydrate is used as pharmaceutical intermediate.
Solubility
Soluble in water and acids. Insoluble in alcohol.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 56378-72-4
Formula molecolare C4H12Mg5O19
Molecular Weight (g/mol) 485.65
Numero MDL MFCD00149784
InChI Key SXKPPTNZIUYFJR-UHFFFAOYSA-D
Sinonimo acmc-20akln, ksc492i1t, magnesium carbonate hydroxide pentahydrate, light, magnesium hydrogen carbonate hydroxide-water 1/1/1/5
IUPAC Name pentamagnesium(2+) pentahydrate dihydroxide tetracarbonate
SMILES O.O.O.O.O.[OH-].[OH-].[Mg++].[Mg++].[Mg++].[Mg++].[Mg++].[O-]C([O-])=O.[O-]C([O-])=O.[O-]C([O-])=O.[O-]C([O-])=O
Specifications
Materiale o nome chimico Magnesium carbonate hydroxide pentahydrate
Nota nome light
Quantità 1000 g
Densità 2.16
Formula lineare 4MgCO3·Mg(OH)2·5H2O
Informazioni di solubilità Soluble in water and acids. Insoluble in alcohol.
Peso formulazione 485.65 (395.58 Anhydrous)
Percent Purity 98%