Learn More
Magnesium Citrate xHydrate, TRC
Codice prodotto. 30480977
Cambia vista
Click to view available options
Quantità:
5 g
Dimensione della confezione:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Quantità unitSize
30480977 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Codice prodotto. 30480977 Fornitore Toronto Research Chemicals N. del fornitore TRCM2457255G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifica
Materiale o nome chimico Magnesium Citrate xHydrate
CAS 960307-84-0
Formula molecolare C6H8O7.3/2Mg.XH2O
Quantità 5 g
InChI Formula InChI=1S/C6H8O7.Mg.H2O/c7-3(8)1-6(13,5(11)12)2-4(9)10;;/h13H,1-2H2,(H,7,8)(H,9,10)(H,11,12);;1H2/q;+2;/p-2
SMILES O.[Mg].[Mg].[Mg].OC(=O)CC(O)(CC(=O)O)C(=O)O.OC(=O)CC(O)(CC(=O)O)C(=O)O
IUPAC Name magnesium 2-(carboxymethyl)-2-hydroxysuccinate hydrate
Molecular Weight (g/mol) 469.13
Peso formulazione 467.9727