Learn More
Interscience™ Magnifying Lens
Product Code. 12628685
Change view
Click to view available options
Magnification:
2x
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Magnification Unit Size
12628685 2x Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 12628685 Supplier Interscience™ Supplier No. 435001
  • ONLINE EXCLUSIVE PRICE
    340.00€
    305.40€ / pezzi 1
    Risparmia 34.60€
    10% Sconto
Spedizione stimata: 09-12-2026
Magnifying lens for close inspection and counting of colonies.
  • Includes magnifying lens and flexible arm for Scan 50, Scan 50 Pro and Scan 100
Specifications
Color Transparent
For Use With (Application) Magnifying Colonies
For Use With (Equipment) Scan 50, 50 Pro, Scan 100
Magnification 2x
Material Glass
Product Type Lens, Magnifying