Learn More
Value Plastics Male Luer with Integral Lock Ring to 200 Series Barb
Sfoglia Tutto Cole-Parmer ProdottiDescrizione
Rotating locks minimize twisting and kinking of tubing during positioning. Tapered cone seals are reliable and uncomplicated. Facilitate single-use configurations that minimize downtime
Specifica
Specifica
| Certificazione/Conformità | Non-DEHP;Non-Phthalate;RoHS |
| Diametro (sistema imperiale) interno | 1/8 in. |
| Materiale | Nylon |
| Ingresso | Male Luer x Hose Barb |
Warning: This product is not approved or intended for, and should not be used for, medical, clinical, surgical or other patient-oriented applications.